Learn More
Cole Parmer CPC (Colder) Quick-Disconnect Hose Barb Body
Standard flow quick-disconnect couplings require a body and an insert of the same flow size. Acetal
Brand: Cole Parmer 06361-51
Additional Details : Weight : 0.01000kg
Description
Standard flow quick-disconnect couplings require a body and an insert of the same flow size. Acetal
Plug insert into body to connect fitting—fitting halves lock into place immediately. To disconnect fitting halves, simply press the thumb latch on the coupling body.Specifications
Quick Disconnect Hose Barb Body | |
1/4 in. | |
1/4 in. | |
1 | |
White |
Yes | |
1/4 in. | |
4.95 cm | |
1/4 in. |