Learn More
Cole-Parmer™ CPC™ DrumQuik PRO System Pro Drum Insert Plug
Product Code. 15467423
Click to view available options
Material:
Polyethylene
Unit Size:
Each
Product Code. 15467423
Brand: Cole-Parmer™ 3160837
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Closed-system design ensures the safety of your plant, the environment and your employees at all phases of chemical handling
  • Easy-to-use, economical solution for chemical dispensing.
  • Durable materials withstand harsh operating conditions.
  • Ideal for 5-gallon jerry cans, 55-gallon drums and IBCs.
  • The DrumQuik PRO system offers a simplified solution to your chemical dispensing needs.
  • Maintains a closed system, protecting your facility, your operating environment, and your employees from the potential dangers of chemical drips, spills and escaping fumes.
  • Simple connections, rugged construction, and chemical-resistant materials handle a wide variety of applications including acids and caustics, agricultural liquids, cleaning agents and sterilants, paints and coatings and food oils and syrups.
  • Base components are a reusable quick-disconnect coupler and a single-use drum insert assembly; both of which are made from environmentally friendly, food-grade polyethylene (PE).
  • Order a convenient dispensing kit which includes all required components, or customize your own system with individual components.
  • Coupler body and plugs mate equally well with all drum inserts listed.
Specifications
Description Buttress Connection
Material Polyethylene
For Use With 5-gallon jerry cans, 55-gallon drums, and IBCs
Flow Rate 10 GPM at less than 2psid (0.14 bar) pressure drop
Maximum Pressure 45 psi
Product Type DrumQuik Pro Drum Insert Plug