Learn More
Cole-Parmer™ CPC™ High-Flow Valved Coupling Insert
Click to view available options
Hose Barb:
1/2 in.
Unit Size:
Pack of 10
Description
- High flow coupling insert.
- USP Class VI-compliant couplings require a fitting body and an insert (order separately).
- Fitting materials are BSE/TSE-free.
- Valves restrict flow when fitting halves are disconnected.
Specifications
Specifications
| Description | Quick-Disconnect x Barb |
| Material | Polysulfone |
| Hose Barb | 1/2 in. |
| Sterilization Method | Autoclave or Gamma |
| Product Type | High-flow coupling insert, valved, polysulfone, 1/2 in. barbed connection |