Learn More
Cole-Parmer™ CPC Miniature Quick-Disconnect Valved Male Hose Barb
Click to view available options
Hose Barb ID:
1/8 in.
Unit Size:
Pack of 10
Description
- CPC Miniature Quick-Disconnect Valved Male Hose Barb
- Standard flow quick-disconnect couplings require a body and an insert of the same flow size
- Plug insert into body to connect fitting
- Fitting halves lock into place immediately
- To disconnect fitting halves, simply press the thumb latch on the coupling body
Specifications
Specifications
| Description | Quick-Disconnect x Barb Connection |
| Material | Polycarbonate |
| Overall Length (Metric) | 2.286 cm |
| Tubing Size | 1/8 in. |
| Outlet Port | 3/32 in. |
| Hose Barb ID | 1/8 in. |
| Product Type | Valved Male Hose Barb |