Learn More
CPC Colder Products Company™ NS4 Series Quick-Disconnect Coupling Bodies
Product Code. p-3850946
Click to view available options
Overall Length (Metric):
58 mm
59.5 mm
Product Type:
Hose Barb Valved In-line Coupling Body
NPT Valved Coupling Body
Unit Size:
Each
Pack of 25
WARNING: Pressure, temperature, chemicals and operating environment can affect the performance of couplings. It is the customer’s responsibility to test the suitability of Colder products in their own application conditions.
Product Code. 11930247
Brand: CPC Colder Products Company™ 3160327
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
WARNING: Pressure, temperature, chemicals and operating environment can affect the performance of couplings. It is the customer’s responsibility to test the suitability of Colder products in their own application conditions.
Manufactured from polypropylene. CPC Colder Products Company™ NS4 Series Quick-Disconnect Coupling Bodies are made from polypropylene. They have a temperature range between 0 ° to 71° and a pressure range up to 120 psi, 8.3 bar.
  • Polypropylene
  • -0°C to 71°C
  • Mounting: Thread
  • Pressure range: Vacuum to 120psi, 8.3 bar
  • Spilling: <0.10cc per disconnect, 0.10cc per connect
Specifications
Material Polypropylene
Overall Length (English) 2.31 in.
Overall Length (Metric) 58 mm
For Use With 6.4mm
Min. Operating Temperature (English) 32°F
Min. Operating Temperature (Metric) 0°C
Thread Size 0.25 in. NPT
Height (Metric) 26.6 mm
Height (English) 1.05 in.
Includes Shut-off valves
Color Gray with Dark Gray Overmold
Maximum Pressure 120 psi
Max. Operating Temperature (English) 160°F
Max. Operating Temperature (Metric) 71°C
Product Type NPT Valved Coupling Body