Learn More
Cole-Parmer™ CPC™ Polypropylene Valved In-Line Flared Fitting
Product Code. 11776128
Click to view available options
Material:
Polypropylene
Unit Size:
Each
Product Code. 11776128
Brand: Cole-Parmer™ 3160317
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Quick-disconnect fittings are practical, economical and disconnect in a snap
  • ChemQuik quick-disconnect couplings require a fitting body and an insert of the same size (order separately).
  • 100% metal-free construction.
  • Single-handed operation.
  • Valves for flow shut-off when disconnected.
  • Make and break tubing connections without separating the tubing from the fitting.
  • Create your own combinations for enhanced flexibility.
  • Easily and intuitively switch lines to decrease downtime and increase efficiency.
  • Reduce spills with valved fittings for additional safety.
  • Select from various materials including plastic quick disconnects.
  • Available with barbed, pipe, compression and more.
Specifications
Description Quick-Disconnect x Flare
Material Polypropylene
Product Type Flared fitting, valved, 1/4 in. OD. (Polypropylene)