Learn More
Cole-Parmer™ CPC™ Quick Disconnect Stainless Steel Sanitary Couplings
Click to view available options
Tubing Size:
3/4 in.
Unit Size:
Each
Description
- Attach these couplings directly to your sanitary hardware assemblies
USP Class IV materials. - Fully sterilizable.
- Both couplings and inserts are available with standard sanitary terminations to attach to your system with a sanitary gasket and tri-clover clamps.
- Choose from stainless steel or polysulfone inserts to mate to polysulfone bodies.
- All models feature USP Class VI platinum-cured silicone gaskets.
- Mix and match these bodies and inserts with the polysulfone quick-disconnect coupling elements for greater flexibility in your process system.
Specifications
Specifications
| Description | Sanitary x Quick-Disconnect |
| Autoclavable | Autoclavable |
| Material | Stainless Steel |
| Tubing Size | 3/4 in. |
| Sterilization Method | Autoclave or Gamma |
| Product Type | Quick-Disconnect Insert, Stainless Steel, Sanitary Connector |