Learn More
Abeba™ Crawler ALU 34511 Shoes
Product Code. 15619917
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. 15619917
Brand: Abeba™ 34511/45
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
ESD dissipative shoe is Sanitized™ treated. In cludes TPU sole with extremly high slip resistance.
  • CE EN ISO 20345:2011 S1 SRC
  • Microfiber with breathable textile inlays
  • Black/Blue
  • Aluminum toe cap
  • Foamed PU midsole
  • Slip resistance TPU outsole sole
  • Replaceable acc Wave insole (article no. 3577)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Reflective material
  • Discontinued model
Specifications
Certifications/Compliance CE EN ISO 20345:2011 S1 SRC
Color Black, Blue
Material Microfiber
Size 45
Sole Features Slip-resistant
Sole Material Polyurethane
Style 34511
Toe Material Aluminum