Learn More
Abeba™ Crawler ALU 34511 Shoes
ESD dissipative shoe is Sanitized™ treated. In cludes TPU sole with extremly high slip resistance.
Brand: Abeba™ 34511/45
Additional Details : Weight : 1.26000kg
Description
- CE EN ISO 20345:2011 S1 SRC
- Microfiber with breathable textile inlays
- Black/Blue
- Aluminum toe cap
- Foamed PU midsole
- Slip resistance TPU outsole sole
- Replaceable acc Wave insole (article no. 3577)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Breathable lining with Silver ions
- Instep area with lacing
- Reflective material
- Discontinued model
Specifications
CE EN ISO 20345:2011 S1 SRC | |
Microfiber | |
Slip-resistant | |
34511 |
Black, Blue | |
45 | |
Polyurethane | |
Aluminum |