Learn More
Abeba™ Crawler ALU 34570 Shoes
ESD dissipative shoe is Sanitized™ treated. In cludes TPU sole with extremly high slip resistance.
Brand:  Abeba™ 34570/40
Additional Details : Weight : 1.23200kg
 View more versions of this product
Product Code. 15659947
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
  • CE EN ISO 20345:2011 S1 SRC
  • Smooth leather with breathable textile inlays
  • White
  • Aluminum toe cap
  • Foamed PU midsole
  • Slip resistance TPU outsole sole
  • Replaceable acc Wave insole (article no. 3577)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Reflective material
  • Discontinued model
  • Scratch-resistant coating in toe and heel area
Specifications
Specifications
CE EN ISO 20345:2011 S1 SRC
Leather
Slip-resistant
34570
White
40
Polyurethane
Aluminum