Learn More
Abeba™ Crawler ALU 34580 Shoes
ESD dissipative shoe is Sanitized™ treated. In cludes TPU sole with extremly high slip resistance.
134.64€
Specifications
Certifications/Compliance CE EN ISO 20345:2011 S1 SRC
Color White
Material Leather
Sole Features Slip-resistant
Sole Material Polyurethane
View More Specs
Products 12
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
15660008
View Documents Promotion Details
Abeba™
34580/36
36 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15670008
View Documents Promotion Details
Abeba™
34580/37
37 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15680008
View Documents Promotion Details
Abeba™
34580/38
38 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15690008
View Documents Promotion Details
Abeba™
34580/39
39 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15600018
View Documents Promotion Details
Abeba™
34580/40
40 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15610018
View Documents Promotion Details
Abeba™
34580/41
41 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15630018
View Documents Promotion Details
Abeba™
34580/43
43 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15640018
View Documents Promotion Details
Abeba™
34580/44
44 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15650018
View Documents Promotion Details
Abeba™
34580/45
45 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15660018
View Documents Promotion Details
Abeba™
34580/46
46 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15670018
View Documents Promotion Details
Abeba™
34580/47
47 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15680018
View Documents Promotion Details
Abeba™
34580/48
48 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
Description
Description
  • CE EN ISO 20345:2011 S1 SRC
  • Smooth leather white/blue
  • Aluminum toe cap
  • Foamed PU midsole
  • Slip resistance TPU outsole sole
  • Replaceable acc Wave insole (article no. 3577)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Discontinued model
Specifications
Specifications
CE EN ISO 20345:2011 S1 SRC
Leather
Polyurethane
Aluminum
White
Slip-resistant
34580