Learn More
Abeba™ Crawler ALU 34581 Shoes
ESD dissipative shoe is Sanitized™ treated. In cludes TPU sole with extremly high slip resistance.
Brand: Abeba™ 34581/39
Additional Details : Weight : 1.24000kg
Description
- CE EN ISO 20345:2011 S1 SRC
- Velours
- Black/Blue
- Aluminum toe cap
- Foamed PU midsole
- Slip resistance TPU outsole sole
- Replaceable acc Wave insole (article no. 3577)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Breathable lining with Silver ions
- Instep area with lacing
Specifications
CE EN ISO 20345:2011 S1 SRC | |
Velour | |
Slip-resistant | |
34581 |
Black, Blue | |
39 | |
Polyurethane | |
Aluminum |