Learn More
Abeba™ Crawler ALU 34581 Shoes
Product Code. 15690018
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. 15690018
Brand: Abeba™ 34581/36
Only null left
Add to basket
Add to basket
ESD dissipative shoe is Sanitized™ treated. In cludes TPU sole with extremly high slip resistance.
  • CE EN ISO 20345:2011 S1 SRC
  • Velours
  • Black/Blue
  • Aluminum toe cap
  • Foamed PU midsole
  • Slip resistance TPU outsole sole
  • Replaceable acc Wave insole (article no. 3577)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Instep area with lacing
Specifications
Certifications/Compliance CE EN ISO 20345:2011 S1 SRC
Color Black, Blue
Material Velour
Size 36
Sole Features Slip-resistant
Sole Material Polyurethane
Style 34581
Toe Material Aluminum