Learn More
Abeba™ Crawler ALU 34883 Shoes
ESD dissipative shoe is Sanitized™ treated. In cludes TPU sole with extremly high slip resistance.
Brand:  Abeba™ 34883/45
Additional Details : Poids : 1.24000kg
 View more versions of this product
Product Code. 15660108
  • 114.00€ / 1 pair
Expédition estimée
05-10-2020
Add to basket
Description
Description
  • CE EN ISO 20345:2011 S1P SRC
  • Smooth leather
  • Black/white
  • Aluminum toe cap
  • Foamed PU midsole
  • Metalfree penetration resistance
  • Slip resistance TPU outsole sole
  • Replaceable acc Wave insole (article no. 3577)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Discontinued model
Specifications
Specifications
CE EN ISO 20345:2011 S1P SRC
Leather
Slip-resistant
34883
Black
45
Polyurethane
Aluminum