Learn More
Abeba™ Crawler ALU 4502 Shoes
Aluminum toe cap provides lightweight toe protection. Sanitized™ treated.
Brand:  Abeba™ 4502/39
Additional Details : Poids : 1.24000kg
 View more versions of this product
Product Code. 15661528
  • 99.10€ / 1 pair
Expédition estimée
28-09-2020
Add to basket
Description
Description
  • CE EN ISO 20345:2011 S2 SRC
  • Microfiber black/red
  • Aluminum toe cap
  • Foamed PU midsole
  • Slip resistance TPU outsole sole
  • Replaceable acc Wave insole (article no. 3577)
  • Sanitized treated
  • Antistatic
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Reflective material
  • Upper one piece
Specifications
Specifications
CE EN ISO 20345:2011 S2 SRC
Microfiber
Slip-resistant
4502
Black
39
Polyurethane
Aluminum