Learn More
Abeba™ Crawler ALU 4570 Shoes
Aluminum toe cap provides lightweight toe protection. Sanitized™ treated.
Brand:  Abeba™ 4570/41
Additional Details : Poids : 1.23200kg
 View more versions of this product
Product Code. 15661608
  • 104.00€ / 1 pair
Expédition estimée
01-10-2020
Add to basket
Description
Description
  • CE EN ISO 20345:2011 S1 SRC
  • Smooth leather with breathable textile inlays
  • White
  • Aluminum toe cap
  • Foamed PU midsole
  • Slip resistance TPU outsole sole
  • Replaceable acc Wave insole (article no. 3577)
  • Sanitized treated
  • Antistatic
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Reflective material
  • Discontinued model
  • Scratch-resistant coating in toe and heel area
Specifications
Specifications
CE EN ISO 20345:2011 S1 SRC
Leather
Slip-resistant
4570
White
41
Polyurethane
Aluminum