Learn More
Abeba™ Crawler ALU 4572 Shoes
Aluminum toe cap provides lightweight toe protection. Sanitized™ treated.
Brand:  Abeba™ 4572/45
Additional Details : Weight : 1.24800kg
 View more versions of this product
Product Code. 15661638
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
  • CE EN ISO 20345:2011 S1 SRC
  • Velours with breathable textile inlays
  • Black/yellow
  • Aluminum toe cap
  • Foamed PU midsole
  • Slip resistance TPU outsole sole
  • Replaceable acc Wave insole (article no. 3577)
  • Sanitized treated
  • Antistatic
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Reflective material
  • EXPIRED size 36 - 40
  • Scratch-resistant coating in toe and heel area
Specifications
Specifications
CE EN ISO 20345:2011 S1 SRC
Velour
Slip-resistant
4572
Black
45
Polyurethane
Aluminum