Learn More
Abeba™ Crawler ALU 4580 Shoes
Aluminum toe cap provides lightweight toe protection. Sanitized™ treated.
Brand:  Abeba™ 4580/45
Additional Details : Poids : 1.23000kg
 View more versions of this product
Product Code. 15651678
  • 105.00€ / 1 pair
Expédition estimée
28-09-2020
Add to basket
Description
Description
  • CE EN ISO 20345:2011 S1 SRC
  • Smooth leather white/blue
  • Aluminum toe cap
  • Foamed PU midsole
  • Slip resistance TPU outsole sole
  • Replaceable acc Wave insole (article no. 3577)
  • Sanitized treated
  • Antistatic
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Discontinued model
Specifications
Specifications
CE EN ISO 20345:2011 S1 SRC
Leather
Slip-resistant
4580
White
45
Polyurethane
Aluminum