Learn More
Abeba™ Crawler ALU 4883 Shoes
Aluminum toe cap provides lightweight toe protection. Sanitized™ treated.
Brand:  Abeba™ 4883/48
Additional Details : Weight : 1.24000kg
 View more versions of this product
Product Code. 15631748
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
  • CE EN ISO 20345:2011 S1P SRC
  • Smooth leather
  • Black/white
  • Aluminum toe cap
  • Foamed PU midsole
  • Metalfree penetration resistance
  • Slip resistance TPU outsole sole
  • Replaceable acc Wave insole (article no. 3577)
  • Sanitized treated
  • Antistatic
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Discontinued model
Specifications
Specifications
CE EN ISO 20345:2011 S1P SRCi>
Leather
Slip-resistant
4883
Black
48
Polyurethane
Aluminum