Learn More
Abeba™ Crawler ALU 4883 Shoes
Aluminum toe cap provides lightweight toe protection. Sanitized™ treated.
Brand: Abeba™ 4883/43
Additional Details : Weight : 1.24000kg
Description
- CE EN ISO 20345:2011 S1P SRC
- Smooth leather
- Black/white
- Aluminum toe cap
- Foamed PU midsole
- Metalfree penetration resistance
- Slip resistance TPU outsole sole
- Replaceable acc Wave insole (article no. 3577)
- Sanitized treated
- Antistatic
- Breathable lining with Silver ions
- Instep area with lacing
- Discontinued model
Specifications
CE EN ISO 20345:2011 S1P SRCi> | |
Leather | |
Slip-resistant | |
4883 |
Black | |
43 | |
Polyurethane | |
Aluminum |