missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ CRBN5-SNAP2-1C-PIP
Description
CRBN5-SNAP2-1C-PIP is a Degrader (PROTAC™) of SNAP-tag™ labeled fusion proteins (Dmax = 75% at 1 μM). CRBN5-SNAP2-1C-PIP recruits cereblon E3 ligase to induce the degradation of SNAP-fusion proteins. PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license. SNAP-tag is a trademark of New England BioLabs, Inc.
Specifications
Specifications
| Chemical Name or Material | N-(4-(((2-Amino-6-chloropyrimidin-4-yl)oxy)methyl)benzyl)-2-(1-(2-(2,6-dioxopiperidin-3-yl)-1,3-dioxoisoindolin-5-yl)piperidin-4-yl)acetamide |
| Quantity | 5 mg |
| Target | Active Degraders |
| Molecular Formula | C32H32ClN7O6 |
| Purity | 0.97 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?