missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Crosshead Stirring Bars

Product Code. 10787531
Click to view available options
Length (Metric):
8 mm
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 10787531

Brand: Kartell™ 0077100

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Special shape for small containers. Particularly effective stirring action.

  • Crosshead, also known as Crown-shaped, stir bars can be ideal for use in a cuvette or test tube
  • They can be used in other containers as well, so are a good option if you need to switch between two container types
  • Suitable for foodstuff
  • Specifications

    For Use With (Application) Stirring
    For Use With (Equipment) Small containers
    Diameter (Metric) 10 mm
    Length (Metric) 8 mm
    Material Magnet, PTFE
    Quantity 10
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.