Learn More
Eppendorf™ CryoCube™ F740h Freezer - 230 V/50 Hz
GREENER_CHOICE
Product Code. 15907075
Click to view available options
Hinge Side:
Left
Right
Shelves:
3
5
Unit Size:
Each
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Regulatory: This product is under eco-contribution tax.
Product Code. 15907075
Brand: Eppendorf™ F740340021
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Regulatory: This product is under eco-contribution tax.
Eppendorf™ CryoCube™ F740h Freezer - 230 V/50 Hz offers secure sample protection and minimized running costs
  • Classic, LED interface
  • Green cooling liquids
  • Air-cooling
  • 230 V/50 Hz
  • Volume: 740 L (26,1 ft3)
  • Consumes 11,8 kWh/day
  • Holds up to 57,600 samples
  • Pull-down time to -80°C: 4 h 10 min
  • Interior: Stainless Steel
  • Exterior: Powder-coated Steel
  • Polyurethane Foam, Vacuum Insulation Panels
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 740 L
Temperature (Setpoint) Pull-down time to -80°C: 4 h 10 min
Inner Doors 3
Hinge Side Left
Insulation Vacuum, Polyurethane Foam
Interior Stainless Steel
Exterior Finish Powder-coated Steel
Shelves 3
Display LED
Dimensions (DxWxH) Exterior 91.5 × 109.9 × 197.3 cm
Dimensions (D x W x H) Interior 62.1 × 86.5 × 139 cm
Chart Recorder No
Setpoint Security Password Protected
Backup Systems Not Available
Electrical Requirements 230 V, 50 Hz
Energy Usage Consumes 11.8 kWh/day
Refrigerant R290 / R170
Shipping Weight (Metric) 357 to 366 kg
Holds Samples (57,600)
Rack Capacity 18 to 30