Learn More
Saint Gobain Life Sciences Tygon™ E-3603 Non-DEHP Tubing
Formulated for resistance to flex fatigue and abrasion and is less permeable than rubber tubing. Tygon™ E-3603 Non-DEHP Tubing handles virtually all inorganic chemicals found in the lab.
Brand:  Saint Gobain Life Sciences ACF00014-C
Additional Details : Weight : 1.26400kg
 View more versions of this product
Product Code. 11750105
  • 244.00€ / 15m
Estimated Shipment: 25-09-2024
to see stock.
Add to basket
Description
Description
  • The new bio-based formulation delivers the same excellent performance
  • Lot-to-lot consistency for reproducible results
  • Non-toxic and non-contaminating
  • Smooth, polished inner wall
  • Slips easily over fittings and grips securely for simple lab set-ups
  • Contains no BPA or phthalates
  • Standard sizes available to hold full vacuum at room temperature
Typical Applications
  • General laboratory
  • Food and beverage
  • Biopharmaceutical
  • Analytical instruments
  • Peristaltic and vacuum pumps
  • Ideal for condensers, incubators, desiccators, gas lines and drain line
Specifications
Specifications
Tubing
0.19 in.
0.44 in.
49.2 ft.
1/8 in.
Crystal Clear
FDA Compliant, NSF, 3-A, USP Class VI, Japan Food Sanitation Law (#370/1959,1935/2004/EC and 10/2011/EU)
4.8 mm
11.20 mm
15 m
3.2 mm
74°C