Learn More
Multiroir Controlec™ Crystal Polystyrene Storage Drawers
Product Code. 11798283
Click to view available options
Capacity (Metric):
1.9 kg
7 kg
Unit Size:
Each
Product Code. 11798283
Brand: Multiroir Controlec™ 11101MUO19TR
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
View the contents of Multiroir Controlec™ Crystal Polystyrene Storage Drawers clearly due to their construction of crystal clear polystyrene. The drawers also feature a slide assembly, with dividers available to order separately.
  • Total visibility
  • Crystal polypropylene transparent;- like glass
  • Slide assembly
  • Drawer separators available separately
Specifications
Capacity (Metric) 1.9 kg
Description Handle type: Slider assembly
Color Transparent
Material Polystyrene
Shape Rectangular
Product Type Drawer
Quantity 1 Each
Dimensions (L x W x H) 168x235x72 mm
Length (Metric) 235 mm
Width (Metric) 168 mm
Height (Metric) 72 mm