Learn More
Cole-Parmer™ CrystalVu™ Luer Fitting with Male Lueur Lock x 1/8 in. Hose Barb
Product Code. 15345046
Click to view available options
Color:
Berry
Lime
Orange
Rose
Yellow
Unit Size:
Pack of 25
Product Code. 15345046
Brand: Cole-Parmer™ 4552033
Only null left
Add to basket
Add to basket
Available in 6 colors for quick and easy identification: berry, rose, yellow, lime, violet and orange
  • Made of polyolefin, a transparent polycarbonate alternative containing no bisphenol-A (BPA).
  • Superior chemical resistance compared to polycarbonate.
  • Sterilize by autoclave, gamma irradiation or ethylene oxide.
  • Meets USP Class VI.
  • Animal-derived ingredient-free.
  • Available in packs of 25.
Specifications
Description Male luer lock
Material Polyolefin
Packaging Quantity 25/Pk.
Color Lime
Product Type Male Luer Lock Fitting