missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CSNK1E (CK1 epsilon) [R178C], GST Tag Recombinant Protein

Product Code. 17138835
Click to view available options
Quantity:
1 mg
10 μg
100 μg
Unit Size:
100µg
10µg
1mg
This item is not returnable. View return policy

Product Code. 17138835

Brand: Invitrogen™ A33721

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This CSNK1E (CK1 epsilon) R178C recombinant human protein (full length) was expressed in insect cells. CSNK1E (CK1 epsilon) R178C recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the Z'-LYTE™ assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Full Length, Recombinant, Active The gene accession number for CSNK1E (CK1 epsilon) [R178C] is NP_001885.1.

CSNK1E serine/threonine kinase is a member of the CK1 family which plays an important role in diverse cell processes, including DNA replication and repair. CSNK1E is a regulator of Yes-associated protein (YAP) transcription coactivator, which is a key regulator of organ size and a candidate human oncogene. CSNK1E is activated by CCK2R and then phosphorylates PKD2 at Ser244. Phosphorylation of PKD2 leads to its nuclear accumulation and efficient phosphorylation of nuclear PKD2 substrates in human gastric cancer cells. CSNK1E can phosphorylate topoisomerase (topo) II alpha at serine-1106 and this regulates the enzyme activity and sensitivity to topo II-targeted drugs.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P49674
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris HCl with 25% glycerol, 0.1mM PMSF, 0.25mM DTT, 0.1mM EDTA, 150mM NaCl, 10mM glutathione and no preservative; pH 7.5
Gene ID (Entrez) 1454
Molecular Weight (g/mol) 73.6 kDa
Name Human CSNK1E (CK1 epsilon) [R178C], GST Tag
pH Range 7.5
Purification Method Purified
Quantity 1 mg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias CK1
Common Name CK1 epsilon (R178C Mutant)
Gene Symbol CSNK1E
KinaseFamily CK1 Kinase Family
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Shipping Condition Dry Ice
Expression System Insect cells
Protein Form Full Length, Recombinant, Active
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.