missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ CST 967

Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
CST 967 is a potent and selective ubiquitin specific protease 7 (USP7) Degrader (PROTAC™; Dmax = 85% at 1μM, DC50 = 17 nM).
Specifications
Specifications
| Chemical Name or Material | 3-(1-(1-(2-(2,6-Dioxopiperidin-3-yl)-1,3-dioxoisoindolin-5-yl)piperidine-4-carbonyl)piperidin-4-yl)-N-(3-((4-hydroxy-1-((R)-3-phenylbutanoyl)piperidin-4-yl)methyl)-4-oxo-3,4-dihydroquinazolin-7-yl)propanamide |
| Quantity | 5 mg |
| Target | Active Degraders |
| Molecular Formula | C51H58N8O9 |
| Purity | 0.96 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction