Learn More
Thermo Scientific™ CTAP
Product Code. p-5384664
Click to view available options
Quantity:
1 mg
2.5 mg
Unit Size:
1mg
2.5mg
Product Code. 15487489
Brand: Thermo Scientific™ J66219.M
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
For cellular and molecular biology applications
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Purification Method HPLC
Content And Storage -30°C to -10°C
Format Solid
Molecular Weight (g/mol) 1104.30
Product Type CTAP
Quantity 1 mg
Regulatory Status RUO - research use only
Sequence D-Phe-Cys-Tyr-D-Trp-Arg-Thr-Pen-Thr-NH2 (Disulfide bridge: Cys2-Pen7)