missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTBP2 Antibody (PCRP-CTBP2-1A9), Alexa Fluor™ 750 - C-terminus, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08549AF750
491.20 EUR valid until 2025-03-28
Use promo code "25306" to get your promotional price.
This item is not returnable.
View return policy
Description
CTBP2 Monoclonal antibody specifically detects CTBP2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Protein Array
Specifications
CTBP2 | |
Monoclonal | |
Alexa Fluor 750 | |
CtBP2, C-terminal binding protein 2, 10ribeye, C-terminal-binding protein 2 | |
Recombinant full-length human CTBP2 protein (Uniprot: P56545) | |
100 μg | |
Cell Cycle and Replication, Wnt Signaling Pathway | |
1488 | |
Store at 4°C in the dark. | |
IgG2b |
Flow Cytometry, Immunofluorescence, Protein Array | |
PCRP-CTBP2-1A9 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction