Learn More
Thermo Scientific™ Culti-Loops™ Klebsiella oxytoca ATCC™ 8724™
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Brand: Thermo Scientific™ R4603009
Additional Details : Weight : 0.40000kg Transport : UN number : 3373 Chem class : 6.2
Description
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
Specifications
872TM* | |
Klebsiella oxytoca | |
Gram-negative Rods | |
5 Loops/Pack |
Klebsiella oxytoca | |
Culti-Loops Quality Control Organism | |
Qualitative QC | |
NRRL B199, [BUCSAV 143, CCM 1901, NCDC 38068, NCIB 8017, NCIB 8521, NRC 474] |
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.