Learn More
Thermo Scientific™ Culti-Loops™ Serratia marcescens ATCC™ 13880™
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Brand:  Thermo Scientific™ R4608002
Additional Details : Weight : 0.40000kg Transport : UN number : 3373 Chem class : 6.2
Product Code. 12905843
  • 245.00€ / Pack of 5
Estimated Shipment: 01-07-2024
to see stock.
Description
Description
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
Specifications
Specifications
13880TM*
Culti-Loops
Gram-negative Rods
5 Loops/Pack
Serratia marcescens ATCC™ 13880™
Culti-Loops Quality Control Organism
Qualitative QC
BS 303 [CDC 813-60, NCIB 9155, NCTC 10211]
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.