Learn More
OHAUS™ Culture Platform
Product Code. 15782479 Shop All Ohaus Products
Click to view available options
Dimensions (L x W):
45.7 x 45.7 cm
45.7 x 61 cm
Unit Size:
Each
Product Code. 15782479
Brand: OHAUS™ 30400066
Only null left
Add to basket
Add to basket
OHAUS™ Culture Platform designed to hold irregular samples or traditional flask clamps and tube racks.
  • A variety of platform options are available to hold irregular samples or traditional flask clamps and tube racks
  • Platforms can increase the amount of usable space for samples
  • In some cases can be stacked to increase throughput and double sample capacity
TRUSTED_SUSTAINABILITY
Specifications
Product Type Culture Platform
Material Stainless Steel
Depth (English) 18.00 in.
Depth (Metric) 45.70 cm
Dimensions (L x W) 45.7 x 45.7 cm
Length (English) 18.00 in.
Length (Metric) 45.70 cm
Width (English) 18.00 in.
Width (Metric) 45.70 cm
Description Culture Platform with Mat, 46 X 46 cm
Includes Mat