Learn More
Value DNA Oligo , Cartridge, Dry, < 24 , 25 nmol
Custom DNA Oligo, 50N, Cartridge, Dry
Brand: Invitrogen A15614
22.87 EUR valid until 2024-06-28
Use promo code "21649" to get your promotional price.
Additional Details : Weight : 0.25000kg
Description
Value Oligos are the most cost-effective and fastest way to order oligonucleotides from us. Simply enter your sequence and then check out. Value Oligos are available at 25 nmol or 50 nmol scales, with a range of purification options to suit your needs.Specifications
Cartridge | |
DNA Oligo, 50 nmol, 50N, Dry, Cartridge | |
Custom DNA Oligo | |
50 nmol |