Learn More
Ambion™ Custom RNA, Stealth, HPLC
Product Code. 10174283
Click to view available options
Product Type:
RNA HPLC In-vivo
Stealth RNA HPLC
Quantity:
20 nmol
5 nmol
500 nmol
Unit Size:
Each
Product Code. 10174283
Brand: Ambion™ 100001863
Custom RNA, Stealth, HPLC
Use this tool for fast and easy online ordering of bench-tested Validated Stealth RNAi™ siRNA, ready-to-order Stealth Select RNAi™ siRNA, bench-tested BLOCK-iT™ Pol II miR Validated miRNA Vector Duopaks, and ready-to-clone BLOCK-iT™ miR RNAi Select.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Room temperature
Description RNA HPLC In-vivo, 5nmol, Lowest Off-target Effects and Highest Potency, Small, Chemically Modified Single-Stranded RNA Molecules Designed to Specifically Inhibit Endogenous miRNAs for the Purpose of Functional Analysis Via Down-Regulation
Product Type RNA HPLC In-vivo
Purity HPLC
Quantity 5 nmol