Learn More
Ambion™ Custom RNA, Stealth, HPLC
Click to view available options
Product Type:
RNA HPLC In-vivo
Stealth RNA HPLC
Quantity:
20 nmol
5 nmol
500 nmol
Unit Size:
Each
Description
Use this tool for fast and easy online ordering of bench-tested Validated Stealth RNAi™ siRNA, ready-to-order Stealth Select RNAi™ siRNA, bench-tested BLOCK-iT™ Pol II miR Validated miRNA Vector Duopaks, and ready-to-clone BLOCK-iT™ miR RNAi Select.
Specifications
Specifications
| Content And Storage | Room temperature |
| Description | Stealth RNA HPLC, 5nmol, Lowest Off-target Effects and Highest Potency, Small, Chemically Modified Single-Stranded RNA Molecules Designed to Specifically Inhibit Endogenous miRNAs for the Purpose of Functional Analysis Via Down-Regulation |
| Product Type | Stealth RNA HPLC |
| Purity | HPLC |
| Quantity | 5 nmol |