missing translation for 'onlineSavingsMsg'
Learn More

Ambion™ Custom Select siRNA, In Vivo Ready

Product Code. 15581471
Click to view available options
Quantity:
1 μmol
250 nmol
Unit Size:
250nmol
Each
This item is not returnable. View return policy

Product Code. 15581471

Brand: Ambion™ 4404007

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Custom Select siRNA, In Vivo Ready

Silencer™ Select custom siRNAs (LNA-modified 21-bp duplex with overhangs) are RNA interference (RNAi) tools ideal for knocking down gene expression in order to study protein function in a wide range of cell types. These siRNAs provide high knock-down efficiencies and specificities.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Room temperature
Description In-vivo Ready Custom Select siRNA, 1μmol, Ideal for Knocking Down Gene Expression in Order to Study Protein Function in a Wide Range of Cell Types, These siRNAs Provide High Knock-down Efficiencies and Specificities
For Use With (Application) Knocking down gene expression in order to study protein function in a wide range of cell types
Product Type In-vivo Ready Custom Select siRNA
Quantity 1 μmol

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.