Learn More
Ambion™ Custom Silencer™ siRNA
Product Code. 15571144
Click to view available options
Quantity:
1 umol
100 nmol
20 nmol
40 nmol
Unit Size:
100nmol
1µmol
20nmol
40nmol
Each
Product Code. 15571144
Brand: Ambion™ AM16210
Custom Silencer™ siRNA
RNA interference (RNAi) is the best way to effectively knock down gene expression to study protein function in a wide range of cell types. Use our GeneAssist™ Custom siRNA Builder to design and order a custom siRNA.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Room temperature
Description Custom siRNA, 20nmol, HPLC Purity, RNAi is the best Way to Effectively Knock Down Gene Expression to Study Protein Function in a Wide Range of Cell Types, Use our GeneAssist Custom siRNA Builder to Design and Order a Custom siRNA
Product Type Custom siRNA
Purity HPLC
Quantity 20 nmol