Learn More
Hach Lange™ Cuvettes
Unique disposable glass cuvette accessory for spectrophotometers/spectral measurements of liquids that are ideal for use in most scientific, research, and other types of laboratories.
Brand:  Hach Lange™ EBK019
Additional Details : Weight : 0.99000kg
 View more versions of this product
Product Code. 10088169
  • 402.00€ / Each
Estimated Shipment: 17-05-2024
to see stock.
Add to basket
Description
Description
  • Disposable glass cuvettes
  • Accessories for spectrophotometers/spectral measurements of liquids
  • Round cuvettes for LICO
  • Made of glass
  • Disposable
  • 11 mm
Specifications
Specifications
3.5 mL
1000 Pack
Cuvette