Learn More
Macherey-Nagel™ Cyano Phases Polygosil™
Brand:  Macherey-Nagel™ 711380.100
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 10576121
  • 1755.00€ / 100g
Estimated Shipment: 10-06-2024
to see stock.
Specifications
5 μm
Liquid Chromatography
Polygosil CN Phases
60 Å
100 g