Learn More
Cyclohexane, BAKER ANALYZED PHOTREX™, J.T. Baker™
Catalog Number p-7138044
Click to view available options
Quantity:
1 L
2.5 L
110-82-7
C6H12
84.16
MFCD00003814
XDTMQSROBMDMFD-UHFFFAOYSA-N
hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
8078
CHEBI:29005
cyclohexane
C1CCCCC1
110-82-7
C6H12
84.16
MFCD00003814
XDTMQSROBMDMFD-UHFFFAOYSA-N
hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
8078
CHEBI:29005
cyclohexane
C1CCCCC1
Does not interfere with the measurement at the specific wavelength being measured. Cyclohexane, BAKER ANALYZED PHOTREX™, J.T. Baker™ is recommended for use in UV, visible, and IR spectrometry applications.
PHOTREX™ solvents are manufactured to minimize lot-to-lot variability and contaminants that can interfere with UV, and in some cases IR, spectra, including residue after evaporation, and acid and base concentrations. Function testing confirms maximum absorbance in selected wavelengths; 50-100% transmittance windows in IR wavelengths are reported.
  • High quality chemicals for laboratory and specialized industrial use
  • Meets the requirements set forth by the American Chemical Society (ACS)
  • Label provides an actual Lot Analysis as a guarantee of purity and consistency
  • Analytical methods used for these determinations are based on standard test procedures described in technical publications
  • Improvements developed by J.T. Baker are employed to achieve a higher degree of reliability
Melting Point 6.4°C
Absorbance Max. 1.00 at 210nm, Max. 0.50 at 220nm, Max. 0.20 at 230nm, Max. 0.08 at 240nm, Max. 0.03 (at 250nm) - 0.01 (at 300-400nm)
Color Colorless
Linear Formula CH2(CH2)4CH2
Solubility Information 0.05g/L in water at 20°C
Formula Weight 84.16g/mol
Percent Purity ≥99.0%
Grade Meets ACS specifications
Density 0.78g/cm3 at 20°C
Boiling Point 81°C
Quantity 1 L
Explosion Limit 1.2 - 8.3 Vol %
Flash Point −18°C
Ignition Point 260°C
Odor Sweet
Packaging Glass
Physical Form Fluid
Residue after Evaporation Max. 0.002%
Water Max. 0.02%
Chemical Name or Material Cyclohexane
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P331-Do NOT induce vomiting.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P370+P378-In case of fire: Use to extinguish.