Learn More
Retsch™ Cyclone Unit
Product Code. p-7161052 Shop All Retsch Products
Click to view available options
For Use With (Equipment):
Cross Beater Mill SK 300
Rotor Beater Mill SR 300
Unit Size:
Each
Product Code. 15924510
Brand: Retsch™ 22.935.0020
Only null left
Add to basket
Add to basket
Retsch™ Cyclone Unit is an optional dust extraction accessory for use with a rotor beater or cross beater mill.
  • Includes a 5 L collecting receptacle
  • To be used with industrial vacuum cleaner
Specifications
Product Type Cyclone Suction Combination Unit
For Use With (Equipment) Rotor Beater Mill SR 300
Includes 5 L collection receptacle