missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Cyclopamine
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Description
- Cyclopamine is a steroid alkaloid that inhibits the Hedgehog pathway at the point of the pathway activator Smoothened
- Cyclopamine binds to the heptahelical bundle of Smoothened, a G protein-coupled receptor, and prevents it from signaling further downstream (Chen et al.)
- Reduces proliferation of rat neural progenitor cells and mouse neurospheres (Lai et al.; Palma and Ruiz i Altaba)
- Reduces proliferation of mouse mammospheres (Liu et al.)
- Promotes differentiation of pancreatic cells from human embryonic stem cells (D'Amour et al.)
- Inhibits the growth of human and mouse medulloblastoma cells, and human glioblastoma cells (Bar et al.), Berman et al.)
Specifications
Specifications
| CAS | 4449-51-8 |
| Synonym | 11-Deoxojervine; Jervine |
| Quantity | 5 mg |
| Molecular Formula | C27H41NO2 |
| Purity | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction