missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Cyclopamine

Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
Inhibitor of hedgehog (Hh) signaling, likely via direct inhibition of Smoothened, the accessory protein to the putative Hh receptor Patched. Anticancer and teratogenic in vivo. Depletes stem-like cancer cells in glioblastoma and blocks tumor engraftment. Can also be used to induce differentiation of human embryonic stem cells (hESCs) into hormone-expressing endocrine cells.
Specifications
Specifications
| CAS | 4449-51-8 |
| Quantity | 1 mg |
| Molecular Formula | C27H41NO2 |
| Formula Weight | 411.63 |
| Purity | >97% |
| Molecular Weight (g/mol) | 411.63 |
| Chemical Name or Material | Cyclopamine |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction