missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclopentanecarbonitrile, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Cyclopentanecarbonitrile is used as a substrate and reagent to investigate the specific activity of nitrile and amide hydrolyzing enzymes isolated from candida guilliermondii cells. It is also used to prepare cyclopentanecarboxamide by using tetrabutylammonium hydrogen sulfate as a catalyst in the presence of reagents such as aqueous hydrogen peroxide and sodium hydroxide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Cyclopentanecarbonitrile is used as a substrate and reagent to investigate the specific activity of nitrile and amide hydrolyzing enzymes isolated from candida guilliermondii cells. It is also used to prepare cyclopentanecarboxamide by using tetrabutylammonium hydrogen sulfate as a catalyst in the presence of reagents such as aqueous hydrogen peroxide and sodium hydroxide.
Solubility
Miscible with toluene.
Notes
Incompatible with strong oxidizing agents and strong bases.
Cyclopentanecarbonitrile is used as a substrate and reagent to investigate the specific activity of nitrile and amide hydrolyzing enzymes isolated from candida guilliermondii cells. It is also used to prepare cyclopentanecarboxamide by using tetrabutylammonium hydrogen sulfate as a catalyst in the presence of reagents such as aqueous hydrogen peroxide and sodium hydroxide.
Solubility
Miscible with toluene.
Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications
Melting Point | -76°C |
Density | 0.912 |
Boiling Point | 67°C to 68°C (10 mmHg) |
Flash Point | 56°C (133°F) |
Molecular Formula | C6H9N |
Refractive Index | 1.441 |
Quantity | 5 g |
UN Number | UN1992 |
Beilstein | 1098717 |
Synonym | cyanocyclopentane, cyclopentyl cyanide, cyclopentane carbonitrile, cyclopentanenitrile, cyclopentyl nitrile, 1-cyanocyclopentane, cyclopentankohlenitrile, acmc-1aqy6, ksc238k0j, cyclopentanecarbonitrile |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'