missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentylacetic Acid, TRC

Product Code. 30349677
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity unitSize
30349677 1 g 1g
1 options
This item is not returnable. View return policy

Product Code. 30349677

Brand: Toronto Research Chemicals TRCC9898451G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cyclopentylacetic Acid
CAS 1123-00-8
Molecular Formula C7 H12 O2
Quantity 1 g
Synonym 2-Cyclopentaneacetic acid, 2-Cyclopentylacetic acid, Cyclopentylacetic acid, NSC, Cyclopentaneacetic acid (6CI, 7CI, 8CI, 9CI, ACI)
InChI Formula InChI=1S/C7H12O2/c8-7(9)5-6-3-1-2-4-6/h6H,1-5H2,(H,8,9)
SMILES OC(=O)CC1CCCC1
IUPAC Name 2-cyclopentylacetic acid
Molecular Weight (g/mol) 128.169
Formula Weight 128.084
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.