Learn More
Agencinox™ Cylindrical Shovel
Product Code. p-8000007
Click to view available options
Capacity (Metric):
500 mL
1 L
2 L
Unit Size:
Each
Product Code. 11771913
Brand: Agencinox™ 915
Only null left
Add to basket
Add to basket
Transfer samples without contamination. Agencinox™ Stainless Steel Micro Spatulas are chemically inert and manufactured with smooth surfaces.
  • 18/10 Stainless steel chemical scoop
  • Available in a variety of sizes
Specifications
Product Type Shovel
Material Stainless Steel
Capacity (Metric) 500 mL
Size 75 mm
Style Cylindrical
Certifications/Compliance AISI 304
Length (Metric) 150 mm