missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cytochalasin B

Product Code. p-3736078 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Packaging:
Glass bottle
Unit Size:
1mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 10174900

Brand: Thermo Scientific Chemicals 228090010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cytochalasin B, CAS# 14930-96-2, also known as phomin, is an organic heterotricyclic compound that is a cell-permeable mycotoxin with potent activity to inhibit cytoplasmic division. | CAS: 14930-96-2 | C29H37NO5 | 479.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Information

• Cytochalasin B is a cell-permeable fungal toxin / mycotoxin considered to be an inhibitor of cytoplasmic division by blocking the formation of contractile microfilaments

Applications

• Cytochalasin B can inhibit platelet aggregation and actin polymerization
• Cytochalasin B is commonly used in studies on cytokinesis due to its ability to act as an inhibitor
• It is used for characterizing some of the polymerization properties of actin
• This compound can also induce DNA fragmentation, inhibit cell division, and inhibit glucose transport, among other molecular processes
• It could be used as a probe for the two hexose-transport systems in rat L6 myoblasts
• Cytochalasin B is commonly used in studies of glucose transporters (GLUT)
• It can be used in in vitro micronucleus assay protocols

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14930-96-2
Molecular Formula C29H37NO5
Molecular Weight (g/mol) 479.62
MDL Number MFCD00077704
InChI Key GBOGMAARMMDZGR-UHFFFAOYNA-N
Synonym cytochalasin b, cytochalasin b from helminthosporium dematioideum
PubChem CID 44634701
IUPAC Name 16-benzyl-5,13-dihydroxy-9,15-dimethyl-14-methylidene-2H,5H,6H,7H,8H,9H,10H,12aH,13H,14H,15H,15aH,16H,17H,18H-oxacyclotetradeca[3,2-e]isoindole-2,18-dione
SMILES CC1C2C(CC3=CC=CC=C3)NC(=O)C22OC(=O)C=CC(O)CCCC(C)CC=CC2C(O)C1=C

Specifications

Melting Point 218.0°C to 223.0°C
Color White
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 1 mg
Merck Index 15, 2787
Specific Rotation + 89.00
Solubility Information Solubility in water: insoluble. Other solubilities: 10.3 mg/mL acetone - 35.4 mg/mL ethanol (24 c), 371 mg/mL dmso - 492 mg/mL dmf (24 c)
Formula Weight 479.6
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Cytochalasin B
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cytochalasin B
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • Reproductive toxicity Category 2
Hazard Statement
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H300-Fatal if swallowed.,Fatal if swallowed.
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P350-IF ON SKIN: Gently wash with plenty of soap and water.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.