missing translation for 'onlineSavingsMsg'
Learn More

Cytokeratin 7 Rabbit, Clone: KRT7/1499R, enQuireBio™

Rabbit Monoclonal Antibody

199.00€ - 510.00€

Specifications

Antigen Cytokeratin 7
Clone KRT7/1499R
Concentration 200μg/mL
Applications Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15920338
View Documents Promotion Details
EnquireBio
3855-RBM9-P1
100 μg
510.00€
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 05-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15910338
View Documents Promotion Details
EnquireBio
3855-RBM9-P0
20 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

Cytokeratin 7
200μg/mL
Monoclonal
Rabbit
10mM PBS with or without 0.05% BSA and 0.05% azide.
3855
Recombinant full-length human KRT7 protein
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
55kDa
KRT7/1499R
Immunohistochemistry
Unconjugated
RUO
NP_011536627
KRT7
IgG κ
Protein A/G Purified
Cytoplasmic
HeLa cells. Carcinoma of ovary, lung, cervix, or breast
Videos
SDS
Documents

Documents

Product Certifications