missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Gene ID (Entrez) | 3855 |
| Protein Family | Cytokeratin 7 |
| Purity | >80%, by SDS-PAGE |
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| Formulation | PBS, 1 M Urea, pH 7.4. |
| Gene Symbol | KRT7 |
| Quantity | 100 μg |
| Research Category | Cell Biology, Cellular Markers |
| Specific Reactivity | This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-55072. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?